ENA249152521 Enamine

2-(3-methylbutoxy)propanoic acid

  • MDL number MFCD09936320
  • Cas Number : 875231-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56540

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56540
Availability details for
Quantity
  • Availability
  • Add to Cart