ENA249414845 Enamine

2-methyl-5-(thiophen-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD00449830
  • Cas Number : 243669-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16319

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16319
Availability details for
Quantity
  • Availability
  • Add to Cart