ENA249414846 Enamine

2-(pyrrolidin-1-yl)-1,3-thiazole

  • MDL number MFCD06358066
  • Cas Number : 24255-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16655
Availability details for
Quantity
  • Availability
  • Add to Cart