ENA249414856 Enamine

N-hydroxy-4-methyl-N-phenylbenzene-1-sulfonamide

  • MDL number MFCD00181235
  • Cas Number : 38557-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-18663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18663
Availability details for
Quantity
  • Availability
  • Add to Cart