ENA249414857 Enamine

2-[(hydroxyimino)methyl]-6-nitrophenol

  • MDL number MFCD31581731
  • Cas Number : 17580-64-2
  • Molecular Weight : 182.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-18684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18684
Availability details for
Quantity
  • Availability
  • Add to Cart