ENA249414878 Enamine

2-chloro-N-[2-(2-methoxyethoxy)-5-(trifluoromethyl)phenyl]propanamide

  • MDL number MFCD13368186
  • Cas Number : 1155012-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28088

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28088
Availability details for
Quantity
  • Availability
  • Add to Cart