ENA249414886 Enamine

(2-methylpropyl)(2-phenylethyl)amine

  • MDL number MFCD08756436
  • Cas Number : 24070-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32552

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32552
Availability details for
Quantity
  • Availability
  • Add to Cart