ENA249414889 Enamine

1-benzyl-3,3-dimethylthiourea

  • MDL number MFCD00087259
  • Cas Number : 2741-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35074

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35074
Availability details for
Quantity
  • Availability
  • Add to Cart