ENA249414893 Enamine

3-(5-amino-1,3,4-oxadiazol-2-yl)benzonitrile

  • MDL number MFCD09804065
  • Cas Number : 1016521-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253...255
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49708

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49708
Availability details for
Quantity
  • Availability
  • Add to Cart