ENA250698322 Enamine

1-phenyl-4-(1H-pyrazol-4-yl)-1H-pyrazole

  • MDL number MFCD14705553
  • Cas Number : 1223598-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25348
Availability details for
Quantity
  • Availability
  • Add to Cart