ENA250698329 Enamine

1,7-diazaspiro[3.5]nonan-2-one hydrochloride

  • MDL number MFCD14705573
  • Cas Number : 1235441-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52210

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52210
Availability details for
Quantity
  • Availability
  • Add to Cart