ENA250698335 Enamine

2,2,2-trifluoroethyl N-[2-(1H-pyrazol-1-yl)pyridin-3-yl]carbamate

  • MDL number MFCD14705587
  • Cas Number : 1221723-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55207
Availability details for
Quantity
  • Availability
  • Add to Cart