ENA250698338 Enamine

4-(2-hydroxyethyl)-1-(4-methylphenyl)-5-oxo-2,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD14705593
  • Cas Number : 1223267-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55873
Availability details for
Quantity
  • Availability
  • Add to Cart