ENA250698340 Enamine

3-(chloromethyl)-1-phenyl-1H,4H,5H,6H-cyclopenta[c]pyrazole

  • MDL number MFCD14705595
  • Cas Number : 123345-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55893

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55893
Availability details for
Quantity
  • Availability
  • Add to Cart