ENA250698342 Enamine

tert-butyl N-[2-(3-aminobenzenesulfonamido)ethyl]carbamate

  • MDL number MFCD14705598
  • Cas Number : 902775-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56040
Availability details for
Quantity
  • Availability
  • Add to Cart