ENA250698344 Enamine

2-(trifluoromethane)sulfinylpyridine-3-carboxylic acid

  • MDL number MFCD14705601
  • Cas Number : 1223025-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56366

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56366
Availability details for
Quantity
  • Availability
  • Add to Cart