ENA250698346 Enamine

2-(3-fluorophenyl)propan-1-amine hydrochloride

  • MDL number MFCD14705603
  • Cas Number : 1221724-60-2
  • Molecular Weight : 189.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56401
Availability details for
Quantity
  • Availability
  • Add to Cart