ENA250698347 Enamine

methyl 5-chloro-2-methoxy-4-{[(2,2,2-trifluoroethoxy)carbonyl]amino}benzoate

  • MDL number MFCD14705604
  • Cas Number : 1223395-43-4
  • Molecular Weight : 341.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56416
Availability details for
Quantity
  • Availability
  • Add to Cart