ENA250698354 Enamine

3-(cyclopentylamino)propanoic acid hydrochloride

  • MDL number MFCD14705615
  • Cas Number : 1221723-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-56477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56477
Availability details for
Quantity
  • Availability
  • Add to Cart