ENA250698357 Enamine

5-(4-bromophenyl)-1,2,4-triazine-3-thiol

  • MDL number MFCD14705619
  • Cas Number : 98592-15-5
  • Molecular Weight : 268.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56548
Availability details for
Quantity
  • Availability
  • Add to Cart