ENA250698360 Enamine

2,2,2-trifluoroethyl N-[3-(2-methyl-1,3-thiazol-4-yl)phenyl]carbamate

  • MDL number MFCD14705622
  • Cas Number : 1221724-56-6
  • Molecular Weight : 316.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56673
Availability details for
Quantity
  • Availability
  • Add to Cart