ENA250698361 Enamine

2,2,2-trifluoroethyl N-[3-(1-methyl-1H-1,2,3,4-tetrazol-5-yl)phenyl]carbamate

  • MDL number MFCD14705623
  • Cas Number : 1221725-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56723
Availability details for
Quantity
  • Availability
  • Add to Cart