ENA250698362 Enamine

2,2,2-trifluoroethyl N-[3-(1-cyclopropyl-1H-1,2,3,4-tetrazol-5-yl)phenyl]carbamate

  • MDL number MFCD14705625
  • Cas Number : 1221724-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56726
Availability details for
Quantity
  • Availability
  • Add to Cart