ENA250698363 Enamine

2,2,2-trifluoroethyl N-(3-{5H,6H,7H-pyrrolo[2,1-c][1,2,4]triazol-3-yl}phenyl)carbamate

  • MDL number MFCD14705626
  • Cas Number : 1221725-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56753
Availability details for
Quantity
  • Availability
  • Add to Cart