ENA250698365 Enamine

2,2,2-trifluoroethyl N-[4-(1H-1,2,4-triazol-1-yl)-2-(trifluoromethyl)phenyl]carbamate

  • MDL number MFCD14705628
  • Cas Number : 1235441-06-1
  • Molecular Weight : 354.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2,2-trifluoroethyl 4-(1H-1,2,4-triazol-1-yl)-2-(trifluoromethyl)phenylcarbamate
Inchi Code 1S/C12H8F6N4O2/c13-11(14,15)4-24-10(23)21-9-2-1-7(22-6-19-5-20-22)3-8(9)12(16,17)18/h1-3,5-6H,4H2,(H,21,23)
Inchi Key KSSYTQRGNVHGCJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56769
Availability details for
Quantity
  • Availability
  • Add to Cart