ENA250698371 Enamine

[(1-benzylpiperidin-2-yl)methyl]urea

  • MDL number MFCD14705636
  • Cas Number : 1221722-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56888
Availability details for
Quantity
  • Availability
  • Add to Cart