ENA250698375 Enamine

3-methyl-5-(naphthalen-2-yl)-1H-1,2,4-triazole hydrobromide

  • MDL number MFCD14705641
  • Cas Number : 1221723-88-1
  • Molecular Weight : 290.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56907
Availability details for
Quantity
  • Availability
  • Add to Cart