ENA250698378 Enamine

4-[ethyl(methyl)amino]benzoic acid hydrochloride

  • MDL number MFCD14705647
  • Cas Number : 1221724-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56958
Availability details for
Quantity
  • Availability
  • Add to Cart