ENA250698379 Enamine

5-[2-fluoro-4-(morpholin-4-yl)phenyl]-1,2,4-triazin-3-amine

  • MDL number MFCD14705648
  • Cas Number : 1235440-01-3
  • Molecular Weight : 275.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-[2-fluoro-4-(4-morpholinyl)phenyl]-1,2,4-triazin-3-ylamine
Inchi Code 1S/C13H14FN5O/c14-11-7-9(19-3-5-20-6-4-19)1-2-10(11)12-8-16-18-13(15)17-12/h1-2,7-8H,3-6H2,(H2,15,17,18)
Inchi Key FMPYBFUPIBMFHW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56970
Availability details for
Quantity
  • Availability
  • Add to Cart