ENA250698380 Enamine

N-(2-hydroxyphenyl)-octahydro-1H-indole-2-carboxamide

  • MDL number MFCD14705649
  • Cas Number : 1251923-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56974
Availability details for
Quantity
  • Availability
  • Add to Cart