ENA250698386 Enamine

3-(chloromethyl)-1-(3-fluorophenyl)-1H-pyrazole

  • MDL number MFCD14705657
  • Cas Number : 1235440-19-3
  • Molecular Weight : 210.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-57086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57086
Availability details for
Quantity
  • Availability
  • Add to Cart