ENA250698389 Enamine

4-(4-fluorophenyl)-1H-imidazole; nitric acid

  • MDL number MFCD14705661
  • Cas Number : 1221724-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57112

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57112
Availability details for
Quantity
  • Availability
  • Add to Cart