ENA250698393 Enamine

methyl 2-[(2Z)-3-(3-chloropropyl)-4-oxo-1,3-thiazolidin-2-ylidene]acetate

  • MDL number MFCD14705666
  • Cas Number : 1235441-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57146

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57146
Availability details for
Quantity
  • Availability
  • Add to Cart