ENA250698397 Enamine

2-[3-(4-chlorophenyl)-1-phenyl-1H-pyrazol-4-yl]ethan-1-amine hydrochloride

  • MDL number MFCD14705671
  • Cas Number : 1235441-46-9
  • Molecular Weight : 334.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57183
Availability details for
Quantity
  • Availability
  • Add to Cart