ENA250698400 Enamine

2-{4-[(4-chlorophenyl)methyl]piperazin-1-yl}pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD14705675
  • Cas Number : 1235439-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57227
Availability details for
Quantity
  • Availability
  • Add to Cart