ENA250698401 Enamine

2-{4-[(4-methoxyphenyl)methyl]piperazin-1-yl}pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD14705676
  • Cas Number : 1221724-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57228
Availability details for
Quantity
  • Availability
  • Add to Cart