ENA250698404 Enamine

1-methyl-5-[(1-methyl-6-oxo-1,6-dihydropyridin-3-yl)amino]-1,2-dihydropyridin-2-one

  • MDL number MFCD14705680
  • Cas Number : 1221725-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57254
Availability details for
Quantity
  • Availability
  • Add to Cart