ENA250698406 Enamine

2-(dodecylsulfanyl)-5-phenyl-3-(prop-2-en-1-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD14705682
  • Cas Number : 1235439-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84...86
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57279

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57279
Availability details for
Quantity
  • Availability
  • Add to Cart