ENA250698408 Enamine

N-(4-hydroxyphenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD14705685
  • Cas Number : 1221724-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57293
Availability details for
Quantity
  • Availability
  • Add to Cart