ENA250698410 Enamine

methyl 1-(4-cyanophenyl)-4-(2-hydroxyethyl)-5-oxo-2,5-dihydro-1H-pyrazole-3-carboxylate

  • MDL number MFCD14705687
  • Cas Number : 1221725-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57304
Availability details for
Quantity
  • Availability
  • Add to Cart