ENA250698412 Enamine

6-(4-fluorophenoxy)pyridine-3-carboximidamide dihydrochloride

  • MDL number MFCD14705696
  • Cas Number : 1221725-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58881
Availability details for
Quantity
  • Availability
  • Add to Cart