ENA250698413 Enamine

3-[(2-methylphenoxy)methyl]benzene-1-carboximidamide hydrochloride

  • MDL number MFCD14705697
  • Cas Number : 1221722-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58882
Availability details for
Quantity
  • Availability
  • Add to Cart