ENA250698414 Enamine

6-(1H-1,3-benzodiazol-1-yl)pyridine-3-carboximidamide dihydrochloride

  • MDL number MFCD14705698
  • Cas Number : 1221725-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 324...326
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58885

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58885
Availability details for
Quantity
  • Availability
  • Add to Cart