ENA250698415 Enamine

3-[(morpholin-4-yl)methyl]benzene-1-carboximidamide dihydrochloride

  • MDL number MFCD14705699
  • Cas Number : 1221723-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58886
Availability details for
Quantity
  • Availability
  • Add to Cart