ENA250698416 Enamine

[1-(4-bromophenyl)ethyl]hydrazine dihydrochloride

  • MDL number MFCD14705701
  • Cas Number : 1235440-27-3
  • Molecular Weight : 288.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58914
Availability details for
Quantity
  • Availability
  • Add to Cart