ENA250698425 Enamine

N-(cyclopropylmethyl)-4-[(cyclopropylmethyl)amino]-2-nitrobenzene-1-sulfonamide

  • MDL number MFCD14705712
  • Cas Number : 1221726-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58991

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58991
Availability details for
Quantity
  • Availability
  • Add to Cart