ENA250698426 Enamine

3-[2-(chloromethyl)phenoxy]pyridine hydrochloride

  • MDL number MFCD14705713
  • Cas Number : 1221726-09-5
  • Molecular Weight : 256.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58993
Availability details for
Quantity
  • Availability
  • Add to Cart