ENA250698427 Enamine

3-(2-aminopropyl)-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD14705714
  • Cas Number : 1221722-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58996
Availability details for
Quantity
  • Availability
  • Add to Cart