ENA250698430 Enamine

N-[2-(thiophen-2-yl)ethyl]piperidine-4-carboxamide hydrochloride

  • MDL number MFCD14705718
  • Cas Number : 1223505-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59035
Availability details for
Quantity
  • Availability
  • Add to Cart