ENA250698433 Enamine

2-amino-N-(1H-pyrazol-4-yl)acetamide dihydrochloride

  • MDL number MFCD14705720
  • Cas Number : 1221726-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59043
Availability details for
Quantity
  • Availability
  • Add to Cart